HIV1 nef Recombinant Protein, Invitrogen™

HIV1 nef Recombinant Protein, RP-4922, from Invitrogen™

Overview
Brand: Invitrogen

Manufacturer Part Number: RP-4922

HIV-1 NEF

UNSPSC: 12352202

Code: LD

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 11574201

Quantity Price
1 £ 594.0 / 20µg
Estimated Shipment date
from Supplier 16-02-2017
Add to basket


Description and Specification

Specification

Quantity 20μg
Formulation 20μg purified protein (0.48mg/mL) in TBS with 50% glycerol, 0.01M beta mercaptoethanol and no preservative; pH 7.5
Conjugate Unlabeled
For Use With (Application) Control, ELISA, Western Blot
Host Species E. Coli
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles
Molecular Weight ∽27
Recombinant Yes
Concentration 0.48mg/mL
Source E. coli
Format Purified
Name HIV1 nef Recombinant Protein
pH Range 7.5
Purification Method Chromatography

HIV-1 Nef is one of the accessory proteins synthesized in the early stage of AIDS virus reproduction and is abundantly found in infected cells. The name derives from its negative factor thought at the beginning but presently it is remarked as the protein which bears a most distinctive biological characteristic of AIDS virus. The protein interacts directly with the signal transduction protein of the host T cell and works effectively on AIDS infection or on long term survival of the infected cells or induces apoptosis of non-infected cells. It is also involved in the endocytosis and degradation of receptor protein of the cell surface such as CD4 and MH4, important for AIDS virus infection.
Host Species: E. Coli
Formulation: 20μg purified protein (0.48mg/mL) in TBS with 50% glycerol, 0.01M beta mercaptoethanol and no preservative; pH 7.5
RP-4922 can be used as a positive control in Western blotting with ABR antibody anti-HIV-1 Nef (PA1-4961, PA1-4962, or PA1-4963), as well as a HIV-1Nef antigen in detection of anti-HIV-1 Nef antibody in Western blotting or ELISA. RP-4922 has a calculated molecular weight of ∽27 kDa. Purity is 90%.