missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human GDF-5 Recombinant Protein
Recombinant Protein
Brand: Invitrogen™ RP8663
Code : LE
Additional Details : Weight : 0.01000kg
Description
Growth and Differentiation Factor-5 (human) Recombinant Protein, RP-8663, from Invitrogen.
Growth/Differentiation Factor 5 (GDF-5) is a growth factor that regulates cell proliferation and differentiation in embryonic and adult tissues. GDF-5 is part of the TGF family of proteins and is closely related to the BMP family of proteins. Recombinant human GDF-5 is a non-glycosylated homodimer, containing two 120 amino acids chains, with a total molecular weight of 27.4 kDa. To enable bacterial expression the N-terminal sequence of Ala-Pro-Leu-Thr was replaced with a Lys. Purity is typically greater than 98% and endotoxin level less than 1 EU/ug. The expected biological activity (ED50) is determined by the ability to induce alkaline phosphatase activity in ATDC5 cells is typically 0.1-1 ug/mL. The specific activity of this protein is 1x10^4 units/mg. Reconstitute using sterile water at 0.1mg/mL and centrifuge prior to opening vial. Gently pipet solution down the sides of the vial. DO NOT VORTEX sample. Store reconstituted material at -20°C and add 0.1% BSA for additional stability. Amino acid sequence: APSATRQGKR PSKNLKARCS RKALHVNFKD MGWDDWIIAP LEYEAFHCEG LCEFPLRSHL EPTNHAVIQT LMNSMDPEST PPTCCVPTRL SPISILFIDS ANNVVYKQYE DMVVESCGCR.Specifications
P43026 | |
Control | |
8200 | |
Human GDF5 | |
E. coli | |
RUO | |
Human GDF5 | |
Human | |
Hu | |
Recombinant | |
Recombinant |
0.1mg/mL | |
0.1%TFA with no preservative | |
∽26.8 | |
50μg | |
-20°C, Avoid Freeze/Thaw Cycles | |
H-GDF-5 | |
GDF5 | |
Unconjugated | |
E. coli | |
Lyophilized | |
purified |