anti-Granulocyte Macrophage-CSF (human) Recombinant, Thermo Scientific™

Granulocyte Macrophage-CSF (Hu) Recombinant protein RP-75574 detects Granulocyte Macrophage-CSF (Hu) in Not Applicable samples, and is validated for Control and WB

Overview
Brand: Invitrogen

Manufacturer Part Number: RP-75572

UNSPSC: 12352203

Code: LD

Additional Details:
Additional Details: Weight: 0.01000kg



Disclaimers: in vitro research only.

Product Code. 11518901

Quantity Price
1 £ 149.0 / 10µg
Estimated Shipment date
from Supplier 25-04-2017
Add to basket


Description and Specification

Specification

Formulation 10μg purified protein in tris HCl with 50% glycerol and no preservative; pH 8
For Use With (Application) Control, Western Blot
Conjugate Unlabeled
Host Species E. Coli
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles
Cross Reactivity Human
Immunogen N/A
Recombinant Yes
Source E. coli
Format Purified
Name Human Granulocyte Macrophage-Colony Stimulating Factor (GM)-CSF Recombinant Protein
Quantity 10μg
pH Range 8
Gene Alias Granulocyte Macrophage Colony Stimulating Factor


Host Species: E. Coli
Formulation: 10μg purified protein in tris HCl with 50% glycerol and no preservative; pH 8
RP-75572 contains purified recombinant human GM-Colony Stimulating Factor protein with an N-terminal hexahistidine tag. RP-75572 may be used as a positive control in many immunological assays.