missing translation for 'onlineSavingsMsg'
Learn More

Granulocyte Macrophage-CSF (human) Recombinant Protein, Invitrogen™

Granulocyte Macrophage-CSF (human) Recombinant Protein, RP-75574, from Invitrogen™

Brand:  Invitrogen RP75574

Code : LD

Additional Details : Weight : 0.01000kg

Product Code. 11538901

  • £2350.46 / 1mg


Estimated Shipment
15-02-2018
Add to basket

Description

Description


Host Species: E. Coli
Formulation: 1mg purified protein in PBS with 50% glycerol and no preservative
RP-75574 contains purified recombinant human GM-Colony Stimulating Factor protein with an N-terminal hexahistidine tag. RP-75574 may be used as a positive control in many immunological assays.
Specifications

Specifications

Control, Western Blot
1mg purified protein in PBS with 50% glycerol and no preservative
1mg
N/A
Granulocyte Macrophage Colony Stimulating Factor
Human
Yes
Purified
Human Granulocyte Macrophage-Colony Stimulating Factor (GM)-CSF Recombinant Protein
E. coli
store at -20°C, Avoid Freeze/Thaw Cycles
Unlabeled
E. coli
Videos