missing translation for 'onlineSavingsMsg'
Learn More

Granulocyte Macrophage-CSF (human) Recombinant Protein, Invitrogen™

Granulocyte Macrophage-CSF (human) Recombinant Protein, RP-75573, from Invitrogen™

Brand:  Invitrogen RP75573

Code : LD

Additional Details : Weight : 0.01000kg

Product Code. 11528901

  • £429.24 / 100µg


Estimated Shipment
27-06-2018
Add to basket

Description

Description


Host Species: E. Coli
Formulation: 100μg purified protein in tris HCl with 50% glycerol and no preservative; pH 8
RP-75573 contains purified recombinant human GM-Colony Stimulating Factor protein with an N-terminal hexahistidine tag. RP-75573 may be used as a positive control in many immunological assays.
Specifications

Specifications

Control, Western Blot
100μg purified protein in tris HCl with 50% glycerol and no preservative; pH 8
8
E. coli
store at -20°C, Avoid Freeze/Thaw Cycles
Unlabeled
E. coli
Purified
Human Granulocyte Macrophage-Colony Stimulating Factor (GM)-CSF Recombinant Protein
100μg
N/A
Granulocyte Macrophage Colony Stimulating Factor
Human
Yes
Videos