missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human GSR, Recombinant Protein
Recombinant Protein
Brand: Invitrogen™ LFP0024
Additional Details : Weight : 0.99000kg
Description
Glutathione Reductase (Hu) Recombinant Protein, LF-P0024, from Invitrogen.
Glutathione reductase (GR) is a member of pyridine nucleotide-disulfide oxidoreductases, which includes the closely related enzymes thioredoxin reductase, lipoamide dehydrogenase, trypanothione reductase and mercuric ion reductase. GR is a cytoplasmic flavoenzyme widely distributed in aerobic organisms. The dimeric protein is composed of two identical subunits, each containing 1 FAD and 1 redox-active disulfide/dithiol as components of the catalytic apparatus. It plays a role in maintaining glutathione (GSH) in its reduced form by catalyzing the reduction of glutathione disulfide (GSSG) : GSSG + NADPH + H+ to 2GSH + NADP+ In most eukaryotic cells, GR maintains the ratio of [GSH]/[GSSG] elevated, and participates in several vital functions such as the detoxification of reactive oxygen species as well as protein and DNA biosynthesis. This protein corresponds to human Glutathione Reductase (44-522 aa with additional 3 amino acids-GSH at N-term), which has a molecular weight of 51.7 kDa. Specific activity: 19.4 U/mg, where one unit will reduce 1 umol of oxidized glutathione per min at pH 7.5 at 25°C. Purity: >95% by SDS-PAGE. After reconstitution, store at -80°C and avoid freeze and thaw cycles.Specifications
P00390 | |
0.02M HEPES with no preservative; pH 7.5 | |
Human GSR | |
0.5mg | |
-20°C | |
Human GSR | |
Human | |
Hu | |
Recombinant | |
Recombinant |
Control | |
2936 | |
7.5 | |
E. coli | |
RUO | |
GSR | |
Unconjugated | |
E. coli | |
Lyophilized | |
purified |
in vitro research only.