missing translation for 'onlineSavingsMsg'
Learn More

Eotaxin (human) Recombinant Protein, Invitrogen™

Eotaxin (human) Recombinant Protein, RP-75537, from Invitrogen™

Brand:  Invitrogen RP75537

Code : LD

Additional Details : Weight : 0.01000kg

Product Code. 11514231

  • £418.56 / 100µg


Estimated Shipment
29-05-2018
Add to basket

Description

Description


Host Species: E. Coli
Formulation: 100μg purified protein in tris with 50% glycerol and no preservative; pH 8
RP-75537 contains purified recombinant human eotaxin protein with an N-terminal hexahistidine tag. RP-75537 may be used as a positive control in many immunological assays.
Specifications

Specifications

Control, Western Blot
100μg purified protein in tris with 50% glycerol and no preservative; pH 8
100μg
N/A
Unlabeled
E. coli
Purified
Human Eotaxin Recombinant Protein
E. coli
store at -20°C, Avoid Freeze/Thaw Cycles
Human
Yes
Videos