missing translation for 'onlineSavingsMsg'
Learn More

Eotaxin (human) Recombinant Protein, Invitrogen™

Eotaxin (human) Recombinant Protein, RP-75536, from Invitrogen™

Brand:  Invitrogen RP75536

Code : LD

Additional Details : Weight : 0.01000kg

Product Code. 11504231

  • £173.70 / 10µg


Estimated Shipment
21-03-2018
Add to basket

Description

Description


Host Species: E. Coli
Formulation: 10μg purified protein in PBS with 50% glycerol and no preservative
RP-75536 contains purified recombinant human eotaxin protein with an N-terminal hexahistidine tag. RP-75536 may be used as a positive control in many immunological assays.
Specifications

Specifications

Control, Western Blot
10μg purified protein in PBS with 50% glycerol and no preservative
10μg
N/A
Unlabeled
E. coli
Purified
Human Eotaxin Recombinant Protein
E. coli
store at -20°C, Avoid Freeze/Thaw Cycles
Human
Yes
Videos