Thermo Scientific™ Pierce™ ECL 2 Western Blotting Substrate

Recommended HRP substrate for fluorescence imagers

Overview
Brand: Thermo Scientific™

Manufacturer Part Number: 80196

Pierce ECL 2 Western Blotting Substrate 100mL kit

UNSPSC: 12352200

Code: A0

Additional Details:
Additional Details: CAS Number: 123-91-1 Weight: 2.49480kg Transport: UN number : 1993 Chem class : 1 Pack group : II



Product Code. 11557910

Quantity Price
1 £ 212.0 / 100mL
EU Stock 36
Add to basket


  • Description and Specification
  • Alternative Products
Description and Specification

Specification

Storage Requirements 2° to 8°C
Kit Contents Detection reagent 1 and 2
Product Type Pierce ECL 2 western blotting Substrate
Sufficient For 10 Mini-gel blots, 1000cm2 of membrane
Quantity 100mL
Alternative Products