Thermo Scientific™ Pierce™ ECL 2 Western Blotting Substrate

Recommended HRP substrate for fluorescence imagers

Overview
Brand: Thermo Scientific™

Manufacturer Part Number: 80197

UNSPSC: 12352200

Code: PN

Additional Details:
Additional Details: Weight: 0.10000kg



Product Code. 12949947

Quantity Price
1 £ 94.75 / 25mL
Estimated Shipment date
from Supplier 07-03-2017
Add to basket


  • Description and Specification
  • Alternative Products
Description and Specification

Specification

Storage Requirements 2° to 8°C
Product Type Pierce ECL 2 western blotting Substrate
Quantity 25mL
Kit Contents Detection reagent 1 and 2
Sufficient For 250cm2 of membrane
Alternative Products