Learn More
Thermo Scientific™ Phire Green Hot Start II PCR Master Mix
A novel hot start DNA Polymerase for routine and high throughput PCR applications requiring speed and specifity.
£176.00 - £742.00
Spezifikation
GC-Rich PCR Performance | Medium |
---|---|
Polymerase | Phire Hot Start II DNA polymerase |
Reaction Speed | Fast |
Product Line | Phire |
Product Type | Hot Start PCR Master Mix |
Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
15301742
|
Thermo Scientific™
F126L |
1000 x 50μL Reactions |
£742.00
1000 reactions |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15391732
|
Thermo Scientific™
F126S |
200 x 50 μL Reactions |
£176.00
200 reactions |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Beschreibung
Phire Hot Start II DNA Polymerase is significantly faster, extremely robust, and also capable of amplifying long DNA fragments with high yields. These features are achieved through advanced protein engineering of the polymerase. It incorporates a unique double-stranded DNA binding domain which allows short extension times (10 to 15 s/kb), improves yields, and increases fidelity 2-fold compared to Taq DNA polymerase.
The hot start modification of Phire Hot Start II DNA Polymerase is based on the same Affibody inactivation technology utilized by Phusion Hot Start II DNA Polymerase. This technology increases the specificity of the PCR reaction with no additional time required for initial activation of the enzyme.
New Phire Green format is a combination of Phire Hot Start II DNA Polymerase and 5X Phire Green Buffer. The buffer includes a density reagent and tracking dyes for direct loading of PCR products on a gel. The colored buffer does not interfere with enzyme performance and is compatible with downstream applications such as DNA sequencing, ligation and restriction digestion.
Thermo Scientific Phire Hot Start II DNA Polymerase is a novel PCR enzyme for routine and high throughput PCR applications. It outperforms every Taq-based hot start polymerase on the market. Phire Hot Start II DNA Polymerase is significantly faster, extremely robust, and also capable of amplifying long DNA fragments with high yields. These features are achieved through advanced protein engineering of the polymerase. It incorporates a unique double-stranded DNA binding domain which allows short extension times (10 to 15 s/kb), improves yields, and increases fidelity 2-fold compared to Taq DNA polymerase.
The hot start modification of Phire Hot Start II DNA Polymerase is based on the same Affibody inactivation technology utilized by Phusion Hot Start II DNA Polymerase. This technology increases the specificity of the PCR reaction with no additional time required for initial activation of the enzyme.
New Phire Green format is a combination of Phire Hot Start II DNA Polymerase and 5X Phire Green Buffer. The buffer includes a density reagent and two tracking dyes for direct loading of PCR products on a gel. The colored buffer does not interfere with enzyme performance and is compatible with downstream applications such as DNA sequencing, ligation and restriction digestion.
Highlights
- Quick hot start - No reactivation step
- Fast enzyme - Amplify four times faster than with hot start Taq
- Robust - Minimal reaction optimization due to high inhibitor tolerance
- High yields - Abundant products due to high efficiency
- Longer PCR products - Amplify significantly longer DNA fragments than with any hot start Taq
- Direct loading on gel with Green Buffer
- Hot Start PCR
- Routine PCR
- Non-high fidelity PCR
- Fast PCR
- High throughput PCR
- Genotyping
Phire Hot Start II DNA Polymerase
- Phire Hot Start II DNA Polymerase
- 5X Phire Reaction Buffer (provides 1.5 mM MgCl2 in the final 1X concentration)
- DMSO
- Phire Hot Start II DNA Polymerase
- 5X Phire Green Reaction Buffer (provides 1.5 mM MgCl2 in the final 1X concentration)
- DMSO
Spezifikation
Medium | |
Fast | |
Hot Start PCR Master Mix | |
2X | |
Built-In Hot Start | |
SuperMix or Master Mix |
Phire Hot Start II DNA polymerase | |
Phire | |
Cloning | |
2 X | |
Blunt | |
DNA |
For Research Use Only. Not for use in diagnostic procedures.