Learn More
Thermo Scientific™ Nunc™Flasks, Non-treated
For cells that do not require Nunclon™ Δ treated surface.
Brand:  Thermo Scientific™ 159926
Additional Details : Weight : 4.60000kg
Product Code. 11377724
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Includes: Flask with cap
Description
Description
Nunc non-treated cell culture flasks are made of high quality optically clear polystyrene for suspension cell culture.
  • White caps for easy identification
  • Angled neck for easy access
  • EasyFlasks with ergonomic filter cap allows constant airflow and minimizes chance of contamination
  • TripleFlask with three parallel layers and footprint of a standard 175cm2 flask same external dimensions
  • Sterilized by gamma irradiation and certified to be non-pyrogenic
Order Info
Extra caps in every case.
Specifications
Specifications
55 mL
Polystyrene
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products