Learn More
Thermo Scientific™ National™ Headspace 20mm Crimp Seals with Septa
National Crimp Seals ensure sample security every time.
Brand: Thermo Scientific™ C4020-32APK
Code : 17
Additional Details : Weight : 0.14580kg
Description
20mm Crimp seals with prefitted septa for headspace vials
- Seals with pre-inserted septa are convenient and minimize contamination due to handling
- Use magnetic seals with CTC/Leap™ Technologies, Gerstel™ and other magnetic transport autosamplers
- PurePak™ PTFE/Silicone seals are packed in glass jars to maintain purity up to the point of use
- Molded polyethylene SepCap™ is suitable for short-term storage or where repeated access to the vial is required
SepCap is for short-term storage. Do not use SepCap at elevated temperatures.
Order Info
Septa and crimp seals also sold separately.
Specifications
Amber | |
Silica Glass | |
Septa |