missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ StarHead™ Magnetic Stir Bars

Greater mixing action at bottom, side, and top of container, even at low stirring speeds

Brand:  Thermo Scientific™ DS6600-0010

Code : 55

Additional Details : Weight : 0.01700kg

 View more versions of this product

Product Code. 10672971

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Specifications

Yes
Test tubes, round bottom or flat-bottom labware
Stir bar
TFE (Tetrafluoroethylene)
Stirbar Star Head® TFE 10mm
8mm
10mm
Videos