Learn More
Thermo Scientific™ Thermo Scientific™ MAb Analysis Ion Exchange and Size Exclusion HPLC Column Kit, 10 μm, 4 mm x 250 mm
Column Phase: Ion Exchange and Size Exclusion
Brand: Thermo Scientific™ 076197
Code : 16
Additional Details : Weight : 1.00000kg
Description
Facilitate monoclonal antibody (mAb) analysis and characterization with the MAb Analysis IEX and SEC Column Kit. This kit includes an ion-exchange (IEX) and size-exclusion (SEC) column. The IEX column is intended for high-resolution and high-efficiency analysis of MAbs and their associated variants, while the SEC column is designed for separation of MAb monomers, aggregates and fragments. This kit is useful for researchers at the beginning of their MAb analysis projects.
Specifications
4 mm | |
10 μm | |
Ion Exchange and Size Exclusion |
250 mm | |
MAbPac | |
Ion Exchange and Size Exclusion |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.