missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Thermo Scientific™ Shelving Options and Accessories for Environmental Chambers and Incubators

Utilize shelving options and accessories to increase the flexibility of your Thermo Scientific™ Environmental Chamber or Incubator.

Brand:  Thermo Scientific™ 224161

Code : 51

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11851130

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Specifications

Reinforced Stainless Steel Shelf and Channels (max. load 68kg)
Stainless Steel
Large capacity reach-in CO2 incubator; ; Environmental chambers 3948, 3907 and 3962
Shelving
Videos

Cat No: 224161 -- Two per unit max., not for shakers; 68.0 kg load maximum with shelf fully inserted and stationary