Learn More
Thermo Scientific™ Convenience Receptacles for Environmental Chambers and Incubators
Provide electricity to an electrical device inside your environmental chamber or incubator.
Brand:  Thermo Scientific™ 190523
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 13459786
  • £194.00 / Each
Call for latest availability
+44 (0)1509 555 500
Add to basket
Specifications
Specifications
RS-485 interface (compatible with 1535 Monitor/Alarm only) Data Output
3962 Environmental Chamber/Incubator
Factory installed
Convenience Receptacles