missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ HyPURITY™ AQUASTAR™ Guard Cartridges

Drop-in guard cartridges offer convenience, economy and effective protection for extending column lifetime

Brand:  Thermo Scientific™ 22505-013001

Code : 16

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10117764

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Requires : Thermo Scientific UNIGUARD direct connection or stand alone holder, sold separately.

Specifications

Specifications

5μm
10mm
3mm
C18
Videos