missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Hypersil™ APS-2 Columns, 3μm and 5μm Particle Size

Versatile amino propyl phase with excellent chromatographic properties in three modes: weak anion exchange, reversed phase and normal phase

Brand:  Thermo Scientific™ 30703-104030

Code : 16

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10458194

  • £379.00 / Each


Estimated Shipment
23-05-2018
Add to basket

Description

Description

  • In reversed phase mode, Hypersil APS-2 columns are excellent for carbohydrate analysis: extra sensitivity for sugars
  • Alternative selectivity to silica when used in normal phase

Specifications

Specifications

4mm
100mm
3μm
APS-2
Videos