Learn More
Thermo Scientific™ High Purity Single Stage Brass Gas Regulators
Recommended for applications where precise flow control is not critical
£618.21 - £664.96
Specifications
Range 0 to 200 psig
View More Specs
Includes: Diffusion-resistant, diaphragm packless outlet valve to maintain system purity
Products 2
Product Code Brand Range Type Price Quantity & Availability  
Product Code Brand Range Type Price Quantity & Availability  
10351505
View Documents Promotion Details
Thermo Scientific™
60180-901
0 to 200 psig CGA 580 with Triple Filter This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
10280005
View Documents Promotion Details
Thermo Scientific™
60180-909
0 to 200 psig CGA 320 with Triple Filter This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description
Description
  • For non-corrosive analytical and process applications
  • Includes a built-in Smart Purifier Base and Cartridge ensuring 99.99999% gas purity
  • Specially designed, convoluted, stainless steel diaphragm provides good regulating performance and maximum purity integrity without the need for a soft seal
  • Capable of withstanding an internal vacuum
  • All regulators supplied with 1/8 in. stainless steel output adaptor
  • Built in check valve allows for cylinder change without contaminating the gas line
Specifications
Specifications
0 to 200 psig