Learn More
Thermo Scientific™ HAAKE™ Viscotester™ iQ Rheometer Application Packages
Ready to use in seconds in any configuration you need
Brand:  Thermo Scientific™ 262-0020
Additional Details : Weight : 12.50000kg
 View more versions of this product
Product Code. 15345123
  • £9489.29 / Each
Estimated Shipment
11-02-2019
Add to basket
Description
Description
Acquire simple viscosity curves or perform complex rheological investigations covering measurements of low viscous liquids up to highly viscous pastes with the Thermo Scientific™ HAAKE™ Viscotester™ rheometer, which can be used as a standalone unit or optionally, be controlled by software. The size and footprint of the HAAKE™ Viscotester™ rheometer make it a portable unit suitable for mobile use or allow it to serve as an important tool in the quality control laboratory.
Application packages are designed to perform a diverse set of rheological tasks and meet a variety of application needs.
  • Advanced design with one-hand lift operation
  • Touchscreen control panel with multilingual, graphical user interface
  • Data quality assessment and QC quality criteria
  • Connect Assist quick coupling and automatic recognition of temperature modules and rotors
  • Liquid temperature module uses a circulator for coaxial cylinders and parallel plates
  • Self-sufficient Peltier temperature module for coaxial cylinders and parallel plates
  • Small dimensioned, coaxial cylinders use less sample and shorter temperature equilibration times to minimize costs per measurement
  • Universal container holder for relative measurements in the original containers
Specifications
Specifications
Particle-containing liquids such as food (e.g. fruit concentrates), mining, and construction samples
HAAKE Viscotester iQ Rheometer, Universal container holder for original containers and Rotor FL22 4B/SS
Application Package for Yield Point Determination
Videos