Thermo Scientific™ GeneJET™ Plasmid Maxiprep Kit

The GeneJET Plasmid Maxiprep Kit is designed for large scale isolation of high quality plasmid DNA from recombinant E. coli cultures.

Overview
Brand: Thermo Scientific™

Manufacturer Part Number: R2231

UNSPSC: 12352200

Code: NEW

Additional Details:
Additional Details: Weight: 0.11000kg Transport: UN number : 1824 Chem class : 1b Pack group : III



Product Code. 15296187

Quantity Price
1 £ 31.13 / 84mL
Estimated Shipment date
from Supplier 14-03-2017
Add to basket


Description and Specification

Specification

For Use With (Equipment) GeneJET™ Endo-free Plasmid Maxiprep Kit
Description Lysis Solution
Storage Requirements Store at room temperature