Thermo Scientific™ GeneJET™ Plasmid Maxiprep Kit

The GeneJET Plasmid Maxiprep Kit is designed for large scale isolation of high quality plasmid DNA from recombinant E. coli cultures.

Overview
Brand: Thermo Scientific™

Manufacturer Part Number: R2271

UNSPSC: 12352200

Code: NEW

Additional Details:
Additional Details: Weight: 0.10000kg



Product Code. 15236197

Quantity Price
1 £ 27.28 / 75mL
Estimated Shipment date
from Supplier 12-04-2017
Add to basket


Description and Specification

Specification

Description Wash Buffer II (conc.)
For Use With (Equipment) GeneJET™ Endo-free Plasmid Maxiprep Kit
Storage Requirements Store at room temperature