Thermo Scientific™ GeneJET™ Plasmid Maxiprep Kit

The GeneJET Plasmid Maxiprep Kit is designed for large scale isolation of high quality plasmid DNA from recombinant E. coli cultures.

Overview
Brand: Thermo Scientific™

Manufacturer Part Number: R2251

UNSPSC: 12352200

Code: NEW

Additional Details:
Additional Details: Weight: 0.02000kg



Product Code. 15216197

Quantity Price
1 £ 33.29 / 12mL
Estimated Shipment date
from Supplier 15-05-2017
Add to basket


Description and Specification

Specification

For Use With (Equipment) GeneJETâ„¢ Endo-free Plasmid Maxiprep Kit
Description Endotoxin Binding Reagent
Storage Requirements Store at room temperature