Thermo Scientific™ GeneJET™ Plasmid Maxiprep Kit

The GeneJET Plasmid Maxiprep Kit is designed for large scale isolation of high quality plasmid DNA from recombinant E. coli cultures.

Overview
Brand: Thermo Scientific™

Manufacturer Part Number: R2241

UNSPSC: 12352200

Code: NEW

Additional Details:
Additional Details: Weight: 0.11000kg Transport: UN number : 2790 Chem class : 1a Pack group : III



Product Code. 15206197

Quantity Price
1 £ 31.65 / 84mL
Estimated Shipment date
from Supplier 12-04-2017
Add to basket


Description and Specification

Specification

For Use With (Equipment) GeneJET™ Endo-free Plasmid Maxiprep Kit
Description Neutralization Solution
Storage Requirements Store at room temperature