Learn More
Thermo Scientific™ Reagents for MagJET™ Plasmid DNA Kit
Brand:  Thermo Scientific™ R2632
Code : 88
Additional Details : Weight : 0.05000kg
 View more versions of this product
Product Code. 13575230
  • £54.41 / 45mL
Estimated Shipment
08-06-2018
Add to basket
Description
Description
The Thermo Scientific™ MagJET™ Plasmid DNA Kit is designed for fast and efficient purification of plasmid DNA from overnight cultures of E. coli cultures. The kit utilizes magnetic bead technology, enabling high yields and robust performance. High binding capacity, uniform particle size, and rapid magnetic response of MagJET magnetic beads makes the technology ideal for high throughput automatic nucleic acid purification, as well as for manual purification by low sample throughput users. The resulting high-quality DNA is free of proteins, nucleases and other contaminants or inhibitors.
Specifications
Specifications
MagJET Plasmid DNA Kit
45mL
Videos