missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Reagents for MagJET™ Plant gDNA Kit

Brand:  Thermo Scientific™ R2501

Code : 88

Additional Details : Weight : 0.07000kg

 View more versions of this product

Product Code. 13555210

  • £33.85 / 16.50mL


Estimated Shipment
02-02-2018
Add to basket

Description

Description

The Thermo Scientific™ MagJET™ Plant Genomic DNA Kit is designed for fast and efficient purification of genomic DNA from a variety of plant species and tissues without phenol/chloroform extraction. The kit utilizes magnetic bead technology, enabling high yields and robust performance. High binding capacity, uniform particle size, and rapid magnetic response of MagJET magnetic beads makes the technology ideal for high throughput automatic nucleic acid purification, as well as for manual purification by low sample throughput users. The resulting high-quality DNA is free of proteins, nucleases and other contaminants or inhibitors.

Specifications

Specifications

MagJET Plant gDNA Kit
16.5mL
Videos