Learn More
Thermo Scientific™ Reagents for MagJET™ gDNA Kit
Brand:  Thermo Scientific™ R2452
Code : 88
Additional Details : Weight : 0.12000kg
 View more versions of this product
Product Code. 13595200
  • £120.62 / 90mL
Estimated Shipment
02-02-2018
Add to basket
Description
Description
The Thermo Scientific™ MagJET™ kits are designed for fast and efficient purification of nucleic acid. The kits utilize paramagnetic bead technology, enabling high yields and robust performance. High binding capacity, uniform particle size, and rapid magnetic response of MagJET magnetic beads makes the technology ideal for high throughput automatic nucleic acid purification, as well as for manual purification by low sample throughput users. The resulting high-quality DNA is free of proteins, nucleases and other contaminants or inhibitors.
Specifications
Specifications
MagJET gDNA Kit
90mL
Videos