Learn More
Thermo Scientific™ Phusion U Hot Start DNA Polymerase
Thermo Scientific Phusion U DNA polymerase is a novel engineered high fidelity enzyme developed using fusion technology.
£159.00 - £706.00
Specifications
Content And Storage | • Phusion U Hot Start DNA Polymerase, 2 U/μL • 5X Phusion HF & GC Buffers • DMSO • 50 mM MgCl2 solution Phusion HF Buffer and Phusion GC Buffers each provide 1.5 mM MgCl2 in the final 1X concentration. Store at -20°C. |
---|---|
GC-Rich PCR Performance | High |
Polymerase | Phusion U Hot Start DNA Polymerase |
Reaction Speed | Fast |
Product Type | Hot Start DNA Polymerase |
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
15220143
|
Thermo Scientific™
F555S |
100 Reactions |
£159.00
100 units |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15230143
|
Thermo Scientific™
F555L |
500 Reactions |
£706.00
500 units |
Estimated Shipment: 01-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Thermo Scientific Phusion U DNA polymerase is a novel engineered high fidelity enzyme developed using fusion technology. Due to a proprietary mutation in the so called dUTP binding pocket of Phusion, Phusion U overcomes an important limitation of proofreading enzymes - it is able to incorporate dUTP and read through uracil present in DNA templates.
In addition to uracil usage possibility, Phusion U features all the superior properties of other Phusion DNA Polymerases - great accuracy, speed, ability to amplify long amplicons up to 20 kb and a high specificity with Affibody based hot start. These features make Phusion U Hot Start DNA Polymerase an excellent choice for such important applications as amplification of bisulphite-converted or damaged DNA as well as use of carryover contamination control.
Phusion U Hot Start DNA Polymerase is also available in the Master Mix and Green formats. Phusion U Hot Start PCR Master Mix (F-533S and F-533L) is convenient 2X mix designed to minimize the number of pipetting steps. Only template and primers need to be added by the user. Phusion U Green format (F-556S and F-556L) is a combination of Phusion U Hot Start DNA Polymerase and 5X Phusion Green Buffers. The buffers include a density reagent and two tracking dyes for direct loading of PCR products on a gel. The colored buffer does not interfere with Phusion U performance and is compatible with downstream applications such as DNA sequencing, ligation and restriction digestion.
Highlights
• Accuracy—high fidelity DNA polymerase (25x Taq)
• Uracil-tolerance—engineered to incorporate dUTP and amplify uracil containing templates
• Specificity—hot start for reduced nonspecific amplification and primer degradation
• Speed—short extension times (15-30 s/kb)
Applications
• Amplification of bisulphite-converted DNA
• Amplification of damaged or aged DNA
• Carry-over contamination control
• Uracil-excision based (USER) cloning methods
Using Phusion DNA Polymerases
Annealing rules for Phusion DNA Polymerases are different from many common DNA polymerases (such as Taq DNA polymerases). For optimal results, use our Tm calculator at www.thermofisher.com/tmcalculator.
Specifications
• Phusion U Hot Start DNA Polymerase, 2 U/μL • 5X Phusion HF & GC Buffers • DMSO • 50 mM MgCl2 solution Phusion HF Buffer and Phusion GC Buffers each provide 1.5 mM MgCl2 in the final 1X concentration. Store at -20°C. |
|
Phusion U Hot Start DNA Polymerase | |
Hot Start DNA Polymerase | |
Hot-start PCR, High-fidelity PCR | |
25X | |
Blunt | |
20 kb or less |
High | |
Fast | |
Dry Ice | |
2 U/μL | |
Built-In Hot Start | |
Standalone |
For Research Use Only. Not for use in diagnostic procedures.