Learn More
Thermo Scientific™ MagJET™ Whole Blood gDNA Kit
Purifies genomic DNA from fresh or frozen whole blood treated with EDTA or citrate, using magnetic bead technology.
Brand:  Thermo Scientific™ K2742
Code : 88
Additional Details : Weight : 1.00000kg Transport : UN number : 1502 Chem class : 1 Pack group : II
 View more versions of this product
Product Code. 13505180
  • £884.45 / Pack of 4
Estimated Shipment
29-03-2018
Add to basket
Specifications
Specifications
PCR and qPCR, FastDigest and conventional restriction digestion, Southern Blotting
Lysis Buffer (60mL), MagJET Magnetic Beads (10.6mL), Proteinase K (8 x 1mL), Binding Buffer (90mL), conc. Wash Buffer 1 (4 x 100mL), conc. Wash Buffer 2 (2 x 40mL), Elution Buffer (3 x 30mL)
4 x 96 Preps
Store Proteinase K solution at room temperature as long as the vial remains sealed. After opening, store at -20deg.C. MagJET Magnetic Beads should be stored at 4deg.C. Other components of the kit should be stored at room temperature (15deg. to 25deg.C).
Videos