Learn More
Thermo Scientific™ MagJET Plasmid DNA Kit
Thermo Scientific MagJET Plasmid DNA Kit, sufficient for 96 preps, is designed for fast and efficient purification of plasmid DNA (up to 4–5 μg/mL) or high copy plasmid DNA from overnight E. coli culture.
Brand: Thermo Scientific™ K2792
369.35 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : 88
Additional Details : Weight : 1.60000kg Transport : UN number : 1824 Chem class : 8 Pack group : III
Description
Thermo Scientific MagJET Plasmid DNA Kit, sufficient for 96 preps, is designed for fast and efficient purification of plasmid DNA (up to 4–5 μg/mL) or high copy plasmid DNA from overnight E. coli culture.
The kit utilizes paramagnetic bead technology enabling high yields and robust performance. High binding capacity, uniform particle size, and rapid magnetic response of MagJET Magnetic Beads makes the technology ideal for high throughput automatic nucleic acid purification, as well as for manual purification for low sample throughput users.
The resulting high quality DNA is free of proteins, nucleases and other contaminants or inhibitors. Purified plasmid DNA can be used in a wide range of downstream applications such as PCR, qPCR and other enzymatic reactions.
Highlights
• High yields—up to 5 μg of plasmid DNA from 1 mL of bacterial culture
• Pure—isolated DNA is immediately ready-to-use in downstream application such as sequencing, transcription, digestion with restriction enzymes or transfection of robust cell lines
• Flexible—automation and manual processing protocols available
• Repeatable results and robust performance
Applications
Fast isolation of high purity plasmid DNA suitable for all conventional molecular biology procedures, including:
• FastDigest or conventional restriction digestion
• Automated fluorescent and radioactive sequencing
• PCR
• In vitro transcription
• Transformation
• Transfection of robust cell lines
Specifications
Kit | |
Bacteria | |
High-throughput Compatible, Not High-throughput Compatible (Manual) | |
MagJET | |
4 x 96 Preps |
Magnetic Bead | |
Real-Time Quantitative PCR (qPCR), Transfection, Cloning, Sequencing, In Vitro Transcription, Transformation, Nucleic Acid Labeling, PCR | |
< 100 μg (Small-Scale) Plasmid DNA | |
Plasmid DNA Kit | |
Plasmid DNA |
For Research Use Only. Not for use in diagnostic procedures.