missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Luminaris HiGreen qPCR Master Mix, fluorescein

Thermo Scientific Luminaris Color HiGreen and Luminaris HiGreen qPCR Master Mixes are universal ready-to-use solutions optimized for qPCR and two-step RT-qPCR.

Brand:  Thermo Scientific™ K0984

Code : 88

Additional Details : Weight : 0.20000kg

 View more versions of this product

Product Code. 13565260

  • £3305.00 / 5000 reactions
Estimated Shipment: 30-04-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Includes:
  • Luminaris HiGreen Fluorescein qPCR Master Mix (2X) containing Hot Start Taq DNA Polymerase, UDG, dNTPs (with dUTP) and SYBR Green I in an optimized PCR buffer. The master mix contains fluorescein passive reference dye for specific real-time platfo

Description

Description

  • UDG in the master mix to prevent carry-over contamination
  • Equal performance of blue and colorless master mixes
  • Versions with premixed ROX or fluorescein for different real-time platforms
  • Standard cycling protocols
  • Excellent qPCR performance
  • Specificity: Hot Start Taq DNA Polymerase and the optimized buffer eliminate non-specific amplification and formation of primer dimers
  • Sensitivity: single copy detection
  • Wide linear range: accurate quantification across 9 orders of magnitude
  • Reproducibility and convenience: ready-to-use 2X master mix
Specifications

Specifications

Contains:
• 4 x 12.5 mL Luminaris HiGreen Fluorescein qPCR Master Mix (2X) containing Hot Start Taq DNA Polymerase, UDG, dNTPs (with dUTP) and SYBR Green I in an optimized PCR buffer; sufficient for 5000 x 20 μL reactions. The master mix contains fluorescein passive reference dye for specific real-time platforms.
• 2 x 30 mL Nuclease-Free Water

Store at -20°C.
High
Taq DNA Polymerase
BioRad iCycler iQ, BioRad MyiQ, BioRad iQ5
qPCR Master Mix
Gene Expression
2 X
DNA, dsDNA
SYBR
qPCR
Standard
Luminaris
5000 Reactions
2X
5000 Reactions
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers