missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Luminaris Color HRM qPCR Master Mix

Luminaris Color HRM Master Mix is a convenient ready-to-use master mix designed for accurate detection of DNA sequence variations using High Resolution Melt analysis method.

Brand:  Thermo Scientific™ K1032BID

Code : NEW

Additional Details : Weight : 0.09000kg

 View more versions of this product

Product Code. 15255136

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

  • High performance on most real-time instruments with HRM capabilities
  • Dual-color system for easy reaction setup
  • Superior discrimination of all four SNP classes
  • High sensitivity - accurate results from 30 copies of genomic DNA
  • Exceptional stability of fully assembled reaction - for high-throughput applications
  • Optimized for PikoReal Real-Time System and software
Specifications

Specifications

Contains:
• 10 x 1.25 mL Luminaris Color HRM Master Mix (2X) containing chemically modified Hot Start Taq DNA Polymerase, EvaGreen™ fluorescent dye, dNTPs and MgCl2 in an optimized PCR buffer with a blue dye; sufficient for 1250 x 20 μL reactions
• 3 x 1.25 mL 40X Yellow Sample Buffer
• 10 x 1.25 mL Nuclease-Free Water

Store at -20°C.
High
Taq DNA Polymerase
SNPs (Unknown or Numerous), Epigenetic Targets
qPCR Master Mix
2X
1250 Reactions
HRM
qPCR
HRM (High Resolution Melting) Analysis, Mutation Scanning
Luminaris
250 Reactions
2 X
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.