Thermo Scientific™ ClaSeek™ Library Preparation Kit, Illumina™ Compatible

Generate Illumina™-compatible next-gen sequencing libraries from low sample input using a one-tube PCR-free method with optimized master mixes

Overview
Brand: Thermo Scientific™

Manufacturer Part Number: K1342

ClaSeek NGS Kit Illumina

UNSPSC: 41116133

Code: NEW

Additional Details:
Additional Details: Weight: 0.05000kg



Product Code. 15215136

Quantity Price
1 £ 1136.0 / 48 reactions
EU Stock 2
Add to basket


Description and Specification

Specification

Storage Requirements Store at -20deg.C before opening.
Shelf Life 1 Year
Quantity 48 reactions
Includes ClaSeek End Conversion Master Mix IL, ClaSeek Ligation Mix IL, nuclease-free water
For Use With (Application) DNA fragment library preparation for downstream use in next generation sequencing workflow.