Thermo Scientific™ ClaSeek™ Library Preparation Kit, Illumina™ Compatible

Generate Illumina™-compatible next-gen sequencing libraries from low sample input using a one-tube PCR-free method with optimized master mixes

Overview
Brand: Thermo Scientific™

Manufacturer Part Number: K1341

UNSPSC: 41116133

Code: NEW

Additional Details:
Additional Details: Weight: 0.05000kg



Product Code. 15205136

Quantity Price
1 £ 357.0 / 12 reactions
EU Stock 1
Add to basket


Description and Specification

Specification

Storage Requirements Store at -20deg.C before opening.
Shelf Life 1 Year
Quantity 12 reactions
For Use With (Application) DNA fragment library preparation for downstream use in next generation sequencing workflow.
Includes ClaSeek End Conversion Master Mix IL, ClaSeek Ligation Mix IL, nuclease-free water