Learn More
Thermo Scientific™ Lysis Buffer for MagJET Viral Kit
The Thermo Scientific™ MagJET™ Viral DNA and RNA Kit is designed for fast and efficient purification of viral nucleic acids from human and animal samples, including plasma, serum, saliva, and urine.
Brand: Thermo Scientific™ R2641
38.50 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : 88
Additional Details : Weight : 0.10000kg Transport : UN number : 1760 Chem class : 8 Pack group : III
Description
The Thermo Scientific™ MagJET™ Viral DNA and RNA Kit is designed for fast and efficient purification of viral nucleic acids from human and animal samples, including plasma, serum, saliva, and urine. The kit utilizes magnetic bead technology, enabling high yields and robust performance. High binding capacity, uniform particle size, and rapid magnetic response of MagJET magnetic beads makes the technology ideal for high throughput automatic nucleic acid purification, as well as for manual purification by low sample throughput users. The resulting high-quality DNA and RNA is free of proteins, nucleases and other contaminants or inhibitors.
Specifications
Lysis Buffer | |
MagJET | |
40 mL |
Kit | |
Lysis Buffer |
For Research Use Only. Not for use in diagnostic procedures.