Learn More
Thermo Scientific™ Buffers for MagJET™ Viral Kit
Brand:  Thermo Scientific™ R2651
Code : 88
Additional Details : Weight : 0.04000kg
 View more versions of this product
Product Code. 13505240
  • £36.02 / 45mL
Estimated Shipment
15-06-2018
Add to basket
Description
Description
The Thermo Scientific™ MagJET™ Viral DNA and RNA Kit is designed for fast and efficient purification of viral nucleic acids from human and animal samples, including plasma, serum, saliva, and urine. The kit utilizes magnetic bead technology, enabling high yields and robust performance. High binding capacity, uniform particle size, and rapid magnetic response of MagJET magnetic beads makes the technology ideal for high throughput automatic nucleic acid purification, as well as for manual purification by low sample throughput users. The resulting high-quality DNA and RNA is free of proteins, nucleases and other contaminants or inhibitors.
Specifications
Specifications
MagJET Viral Kit
45mL
Videos