Learn More
Thermo Scientific™ Buffers for MagJET™ gDNA Kit
Brand:  Thermo Scientific™ R2442
Code : 88
Additional Details : Weight : 0.14000kg
 View more versions of this product
Product Code. 13565200
  • £120.68 / 135mL
Estimated Shipment
10-07-2018
Add to basket
Description
Description
The Thermo Scientific™ MagJET™ Genomic DNA Kit is designed for fast and efficient purification of genomic DNA from tissue and cell cultures, bacteria and yeast, as well as from human body samples, such as buccal and urogenital swabs, urine, saliva and hair follicles. The kit utilizes paramagnetic bead technology, enabling high yields and robust performance. High binding capacity, uniform particle size, and rapid magnetic response of MagJET magnetic beads makes the technology ideal for high throughput automatic nucleic acid purification, as well as for manual purification by low sample throughput users. The resulting high-quality DNA is free of proteins, nucleases and other contaminants or inhibitors.
Specifications
Specifications
MagJET gDNA Kit
135mL
Videos