Learn More
Thermo Scientific™ Buffers for MagJET™ Blood gDNA Kit
Brand:  Thermo Scientific™ R2342
Code : 88
Additional Details : Weight : 0.04000kg
 View more versions of this product
Product Code. 13565190
  • £117.58 / 60mL
Estimated Shipment
05-02-2018
Add to basket
Description
Description
The Thermo Scientific™ MagJET™ Whole Blood Genomic DNA Kit is designed for fast and efficient purification of genomic DNA from fresh or frozen whole blood treated with EDTA or citrate, as well as blood fractions. The kit utilizes paramagnetic bead technology, enabling high yields and robust performance. High binding capacity, uniform particle size, and rapid magnetic response of MagJET magnetic beads makes the technology ideal for high throughput automatic nucleic acid purification, as well as for manual purification by low sample throughput users. The resulting high-quality DNA is free of proteins, nucleases and other contaminants or inhibitors.
Specifications
Specifications
MagJET Blood gDNA Kit
60mL
Videos