missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ EPD Mk2, G and N2 Readers

To complement the Thermo Scientific EP DMk2, G and N2. The basic IrDA reader, is ideal for office environments and mobile applications

Brand:  Thermo Scientific™ EPD/1/62570/000

Code : 50

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10186354

  • £234.43 / Each


Estimated Shipment
20-02-2018
Add to basket

Specifications

Specifications

Serial RS232 or USB 1.1 and 2.0 compatible interface direct to PC
USB interface
Basic Irda Reader
50g
For office environments and mobile applications
1m
EasyEPD2 and EasyIssue software
Videos
Documentation