Learn More
Thermo Scientific™ Dionex™ PEEK Universal Fitting for Switchos
Dionex PEEK fitting is universal for Switchos.
Brand: Thermo Scientific™ 164013
Code : 16
Additional Details : Weight : 0.10000kg
Description
- INERT
- Long hex nut and ferrule with groove
- I.D.: 1/16 in.
Specifications
Long Hex Nut And Ferrule With Groove | |
Switchos | |
PEEK | |
nanoLC |