Learn More
Thermo Scientific™ Cheminert Model C2 Microbore Injector
Can be used as an injector or switching valve
Brand:  Thermo Scientific™ CSL2
Code : 16
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 11573960
  • £29.55 / Each
Estimated Shipment
13-06-2018
Add to basket
Description
Description
  • 1/16 in. fittings
  • 0.010 in. ports
  • Available in 6-port or 10-port configurations
  • Available with manual or microelectric actuator
Specifications
Specifications
2μL
Videos