missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ BioBasic™ SEC 60A HPLC Columns
Outstanding size separation of water soluble compounds
£500.82 - £527.34
Specifications
Column Format | Analytical Column |
---|---|
Length (Metric) | 150 mm |
Particle Size | 5 μm |
Pore Size | 60 Å |
Stationary Phase | Proprietary |
Product Code | Brand | Diameter (Metric) | Length (Metric) | Stationary Phase | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Length (Metric) | Stationary Phase | Price | Quantity & Availability | |||
11512811
|
Thermo Scientific™
73305-152130 |
2.1 mm | 150 mm | Proprietary |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11532811
|
Thermo Scientific™
73305-154630 |
4.6 mm | 150 mm | Proprietary |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
- Covers separation of analytes over a wide molecular weight range
- Long column life and high column efficiencies
- Simple mechanism of interaction based on molecular size and shape
- Excellent for sample cleanup
- Straghtforward method development, simple mobile phases
- 60Å pore size
- Particle size: 5μm
Order Info
For information on bulk quantities, please inquire.
Specifications
Analytical Column | |
5 μm | |
Proprietary | |
Silica | |
Size Exclusion and Gel Permeation |
150 mm | |
60 Å | |
BioBasic | |
Mixed Mode |