missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ BioBasic™ SEC 300A HPLC Columns
Outstanding size separation of water soluble compounds
Brand: Thermo Scientific™ 73505-254630
Code : A0
Additional Details : Weight : 1.47420kg
Description
- Covers separation of analytes over a wide molecular weight range
- Long column life and high column efficiencies
- Simple mechanism of interaction based on molecular size and shape
- Excellent for sample cleanup
- Straghtforward method development, simple mobile phases
- 300Å pore size
Order Info
For information on bulk quantities, please inquire.
Specifications
Analytical Column | |
250 mm | |
300 Å | |
BioBasic | |
Mixed Mode |
4.6 mm | |
5 μm | |
Proprietary | |
Silica | |
Size Exclusion and Gel Permeation |