Learn More
Thermo Scientific™ BetaSil™ Silica Javelin™ Guard Columns
Economically extends HPLC column lifetime
Brand: Thermo Scientific™ 70005-012106
Code : A0
Additional Details : Weight : 0.10000kg
Description
Thermo Scientific™ BETASIL™ Silica HPLC columns are based on a highly pure, 100Å silica designed for small molecule analysis. BETASIL Silica columns are used for normal phase separations. These non-bonded silica columns are a powerful and efficient tool for separating non-polar and moderately polar compounds.
- 100Å high purity silica
- Used for the separation of non-polar and moderately polar compounds.
- Direct-connection, fingertight design for convenience and maximum efficiency
- Patented design requires no holder, allowing quick and easy replacement
Features of Betasil Silica Solid Phase
- 100Å silica
- High surface area with high bonded phase coverage
Specifications
5 μm | |
10 mm | |
2.1 mm | |
BetaSil |