missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ BetaSil™ Silica Columns, 5μm Particle Size

High surface-area silica columns

Brand:  Thermo Scientific™ 70005-104030

Code : 16

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10587815

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description

Description

  • 100Å silica

Specifications

Specifications

4mm
Silica
Silica 100
100mm
5μm
Videos