missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ BetaBasic™ Phenyl Columns, 5μm Particle Size
BetaBasic columns are highly efficient and reproducible for small molecules, peptides and protein digests
£522.24 - £706.52
Specifications
Film Thickness | 5 μm |
---|---|
For Use With | LC/MS Separation |
Particle Shape | Spherical |
Particle Size | 5 μm |
Pore Size | 150 Å |
Product Code | Brand | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | ||||
10231145
|
Thermo Scientific™
71805-254630 |
4.6 mm | 250 mm |
£706.52
Each |
In Stock
Log in to see stock. |
||||
10794556
|
Thermo Scientific™
71805-154630 |
4.6 mm | 150 mm |
£673.80
Each |
Estimated Shipment: 16-04-2024
Log in to see stock. |
||||
10408004
|
Thermo Scientific™
71805-252130 |
2.1 mm | 250 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- 150Å pore size for small molecules, peptides and protein digests
- Superb pH stability
Order Info
Other column dimensions are also available. Please call Customer Service for more information.
Specifications
5 μm | |
Spherical | |
150 Å | |
L11 | |
Silica | |
Reversed Phase | |
Reversed Phase |
LC/MS Separation | |
5 μm | |
200 m²/g | |
Phenyl (Ph) | |
7% | |
Yes |