missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ BetaBasic™ Phenyl Columns, 3μm Particle Size
BetaBasic columns are highly efficient and reproducible for small molecules, peptides and protein digests
Brand: Thermo Scientific™ 71803-154630
Code : 16
Additional Details : Weight : 0.10000kg
Description
- 150Å pore size for small molecules, peptides and protein digests
- Superb pH stability
Order Info
Other column dimensions are also available. Please call Customer Service for more information.
Specifications
4.6 mm | |
150 mm | |
3 μm | |
200 m²/g | |
Phenyl (Ph) | |
7% | |
Yes | |
HPLC Column |
3 μm | |
Spherical | |
150 Å | |
L11 | |
Silica | |
Reversed Phase | |
Reversed Phase | |
HPLC |