missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ BetaBasic™ Phenyl Columns, 3μm Particle Size
BetaBasic columns are highly efficient and reproducible for small molecules, peptides and protein digests
£453.00 - £542.64
Specifications
Film Thickness | 3 μm |
---|---|
Particle Shape | Spherical |
Particle Size | 3 μm |
Pore Size | 150 Å |
Surface Area | 200 m²/g |
Product Code | Brand | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | ||||
10681296
|
Thermo Scientific™
71803-154630 |
4.6 mm | 150 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10336524
|
Thermo Scientific™
71803-152130 |
2.1 mm | 150 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- 150Å pore size for small molecules, peptides and protein digests
- Superb pH stability
Order Info
Other column dimensions are also available. Please call Customer Service for more information.
Specifications
3 μm | |
3 μm | |
200 m²/g | |
Phenyl (Ph) | |
7% | |
Yes | |
HPLC Column |
Spherical | |
150 Å | |
L11 | |
Silica | |
Reversed Phase | |
Reversed Phase | |
HPLC |