missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ BetaBasic™ CN Columns, 3 and 5μm Particle Size
Highly efficient and reproducible
£384.93
Specifications
Particle Size | 3 μm |
---|---|
pH | 2 to 9 |
Pore Size | 150 Å |
Surface Area | 200 m²/g |
USP Type | L10 |
Product Code | Brand | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | ||||
10448384
|
Thermo Scientific™
71703-052130 |
2.1 mm | 50 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Pore size suitable for small molecules, peptides and protein digests
- Superb pH stability
Order Info
Other column dimensions are also available. Please call Customer Service for more information.
Specifications
3 μm | |
150 Å | |
L10 | |
Silica | |
Mixed Mode | |
Normal and Reversed Phase |
2 to 9 | |
200 m²/g | |
Cyano (CN) | |
5% | |
Yes |